Learn More
Medchemexpress LLC NBD-14189 | 2234273-72-2 | 99.8% | 428.40 | 100 MG
SDP
Catalog No. 5000349450
Encompass_Preferred
NBD-14189 is a potent HIV-1 entry antagonist, binding to HIV-1 gp120 and demonstrating potent antiviral activity against the HIV-1HXB2 pseudovirus. It shows anti-HIV-1 activity in both single-cycle and multi-cycle assays.
  • Potent HIV-1 entry antagonist
  • Binds to HIV-1 gp120
  • Exhibits potent antiviral activity (EC50 < 200 nM)
  • Prevents HIV-1 mediated cell-cell fusion
  • Shows weak or no inhibition of the hERG current
Catalog No. 50-003-49450 Supplier Medchemexpress LLC Supplier No. HY139985100MG
NBD-14189 is a potent HIV-1 entry antagonist, binding to HIV-1 gp120 and demonstrating potent antiviral activity against the HIV-1HXB2 pseudovirus. It shows anti-HIV-1 activity in both single-cycle and multi-cycle assays.
  • Potent HIV-1 entry antagonist
  • Binds to HIV-1 gp120
  • Exhibits potent antiviral activity (EC50 < 200 nM)
  • Prevents HIV-1 mediated cell-cell fusion
  • Shows weak or no inhibition of the hERG current