Learn More
Medchemexpress LLC NBD-14189 | 2234273-72-2 | 99.8% | 428.40 | 50 MG
SDP
Catalog No. 5000350506
Encompass_Preferred
NBD-14189 is a potent HIV-1 entry antagonist with an IC50 of 89 nM against the HIV-1HXB2 pseudovirus. It binds to HIV-1 gp120 and demonstrates potent antiviral activity (EC50 < 200 nM). It is active against HIV-1 in single-cycle (TZM-bl cells) and multi-cycle (MT-2 cells) assays.
  • Potent HIV-1 entry antagonist with an IC50 of 89 nM
  • Binds to HIV-1 gp120
  • Demonstrates potent antiviral activity (EC50 < 200 nM)
  • Prevents HIV-1 mediated cell-cell fusion
Catalog No. 50-003-50506 Supplier Medchemexpress LLC Supplier No. HY13998550MG
NBD-14189 is a potent HIV-1 entry antagonist with an IC50 of 89 nM against the HIV-1HXB2 pseudovirus. It binds to HIV-1 gp120 and demonstrates potent antiviral activity (EC50 < 200 nM). It is active against HIV-1 in single-cycle (TZM-bl cells) and multi-cycle (MT-2 cells) assays.
  • Potent HIV-1 entry antagonist with an IC50 of 89 nM
  • Binds to HIV-1 gp120
  • Demonstrates potent antiviral activity (EC50 < 200 nM)
  • Prevents HIV-1 mediated cell-cell fusion