Learn More
Medchemexpress LLC NBD-14189 | 2234273-72-2 | 99.8% | 428.40 | C18H16F4N4O2S | 5 MG
SDP
Catalog No. 5000222834
Encompass_Preferred
NBD-14189 is a small-molecule HIV-1 entry antagonist that binds the viral gp120 protein and exhibits potent antiviral activity in cell-based assays. It is provided as a high-purity research reagent available as solids and as a DMSO solution, with characterized activity in single- and multi-cycle assays.
  • Binds HIV-1 gp120 to inhibit viral entry.
  • Potent antiviral activity (IC50 ≈ 89 nM; EC50 <200 nM).
  • Characterized in single-cycle (0.089 μM) and multi-cycle (0.18 μM) assays.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • High purity (99.84%) suitable for research use.
  • Storage recommendations provided for solids and solutions.
Catalog No. 50-002-22834 Supplier Medchemexpress LLC Supplier No. HY1399855MG
NBD-14189 is a small-molecule HIV-1 entry antagonist that binds the viral gp120 protein and exhibits potent antiviral activity in cell-based assays. It is provided as a high-purity research reagent available as solids and as a DMSO solution, with characterized activity in single- and multi-cycle assays.
  • Binds HIV-1 gp120 to inhibit viral entry.
  • Potent antiviral activity (IC50 ≈ 89 nM; EC50 <200 nM).
  • Characterized in single-cycle (0.089 μM) and multi-cycle (0.18 μM) assays.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • High purity (99.84%) suitable for research use.
  • Storage recommendations provided for solids and solutions.