Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NBD-14189 | 2234273-72-2 | 99.8% | C18H16F4N4O2S | 10MG
SDP

Catalog No. 5000260359
Encompass_Preferred

NBD-14189 is a small-molecule HIV-1 entry antagonist that binds the viral gp120 envelope protein and blocks virus entry. It shows potent activity in cell-based assays (IC50 ≈ 89 nM against HIV-1 HXB2 pseudovirus; antiviral EC50 <200 nM) and is supplied as a high-purity research compound for in vitro studies.

  • Potent gp120-directed entry antagonist with IC50 ≈ 89 nM.
  • Demonstrated antiviral activity in cell assays (EC50 <200 nM).
  • High purity suitable for research use (≈99.8%).
  • Supplied as solid and as 10 mM solution in DMSO for screening.
  • Molecular formula C18H16F4N4O2S and molecular weight 428.40 g·mol⁻1.
  • Recommended storage: 4°C sealed; in solvent store at -80°C for long term.

Catalog No. 50-002-60359 Supplier Medchemexpress LLC Supplier No. HY13998510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NBD-14189 is a small-molecule HIV-1 entry antagonist that binds the viral gp120 envelope protein and blocks virus entry. It shows potent activity in cell-based assays (IC50 ≈ 89 nM against HIV-1 HXB2 pseudovirus; antiviral EC50 <200 nM) and is supplied as a high-purity research compound for in vitro studies.

  • Potent gp120-directed entry antagonist with IC50 ≈ 89 nM.
  • Demonstrated antiviral activity in cell assays (EC50 <200 nM).
  • High purity suitable for research use (≈99.8%).
  • Supplied as solid and as 10 mM solution in DMSO for screening.
  • Molecular formula C18H16F4N4O2S and molecular weight 428.40 g·mol⁻1.
  • Recommended storage: 4°C sealed; in solvent store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.