Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nbd-14270 | 2411819-82-2 | 99.5% | C18H18F3N5O2S | 10MG
SDP

Catalog No. 5000260885
Encompass_Preferred

NBD-14270 is a pyridine-analog small molecule HIV-1 entry antagonist that binds gp120 and inhibits viral entry. Supplied for research use as a high-purity solid, it shows reported antiviral potency with low cytotoxicity.

  • Purity 99.53% for consistent experimental results.
  • High in vitro potency (IC50 ≈180 nM) against HIV-1 Env-pseudotyped viruses.
  • Low cytotoxicity (CC50 >100 μM) supporting cellular assays.
  • Soluble in DMSO (100 mg/mL) for ready stock solutions.
  • Solid, yellow to brown appearance for easy handling and storage.
  • Recommended storage: sealed, away from moisture and light; in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-60885 Supplier Medchemexpress LLC Supplier No. HY13998910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NBD-14270 is a pyridine-analog small molecule HIV-1 entry antagonist that binds gp120 and inhibits viral entry. Supplied for research use as a high-purity solid, it shows reported antiviral potency with low cytotoxicity.

  • Purity 99.53% for consistent experimental results.
  • High in vitro potency (IC50 ≈180 nM) against HIV-1 Env-pseudotyped viruses.
  • Low cytotoxicity (CC50 >100 μM) supporting cellular assays.
  • Soluble in DMSO (100 mg/mL) for ready stock solutions.
  • Solid, yellow to brown appearance for easy handling and storage.
  • Recommended storage: sealed, away from moisture and light; in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.