Learn More
Medchemexpress LLC NBD-14270 | 2411819-82-2 | 99.53% | 425.43 | 1 ML
SDP
Catalog No. 5000350268
Encompass_Preferred
NBD-14270, a pyridine analogue, is a potent HIV-1 entry antagonist with an IC50 of 180 nM against 50 HIV-1 Env-pseudotyped viruses. It binds to HIV-1 gp120 and demonstrates potent antiviral activity, with low cytotoxicity (CC50>100 μM).
  • Potent HIV-1 entry antagonist
  • IC50 of 180 nM against 50 HIV-1 Env-pseudotyped viruses
  • Binds to HIV-1 gp120
  • Demonstrates potent antiviral activity
  • Low cytotoxicity (CC50>100 μM)
Catalog No. 50-003-50268 Supplier Medchemexpress LLC Supplier No. HY13998910MM/1ML
NBD-14270, a pyridine analogue, is a potent HIV-1 entry antagonist with an IC50 of 180 nM against 50 HIV-1 Env-pseudotyped viruses. It binds to HIV-1 gp120 and demonstrates potent antiviral activity, with low cytotoxicity (CC50>100 μM).
  • Potent HIV-1 entry antagonist
  • IC50 of 180 nM against 50 HIV-1 Env-pseudotyped viruses
  • Binds to HIV-1 gp120
  • Demonstrates potent antiviral activity
  • Low cytotoxicity (CC50>100 μM)