Learn More
Medchemexpress LLC Nbd-556 | 333353-44-9 | 99.5% | C17H24ClN3O2 | 100 MG
SDP
Catalog No. 5000434183
Encompass_Preferred
NBD-556 is a CD4 mimetic and a potent HIV-1 entry inhibitor that blocks the gp120-CD4 interaction. It shows potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Inhibits HIV-1 entry by blocking gp120-CD4 interaction.
  • Shows potent cell fusion and virus-cell fusion inhibitory activity.
  • Inhibits HIV-1 envelope-mediated virus-cell and cell-cell fusion.
  • Inhibits infection of MT-2 cells by laboratory-adapted HIV-1 IIIB, MN, and V32 strains.
  • Inhibits CD4-dependent virus in a dose-dependent manner.
Catalog No. 50-004-34183 Supplier Medchemexpress LLC Supplier No. HY76648100MG
NBD-556 is a CD4 mimetic and a potent HIV-1 entry inhibitor that blocks the gp120-CD4 interaction. It shows potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Inhibits HIV-1 entry by blocking gp120-CD4 interaction.
  • Shows potent cell fusion and virus-cell fusion inhibitory activity.
  • Inhibits HIV-1 envelope-mediated virus-cell and cell-cell fusion.
  • Inhibits infection of MT-2 cells by laboratory-adapted HIV-1 IIIB, MN, and V32 strains.
  • Inhibits CD4-dependent virus in a dose-dependent manner.