Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NBD-556 | 333353-44-9 | 99.6% | 337.84 g·mol-1 | C17H24ClN3O2 | 10 MG
SDP

Catalog No. 5000298491
Encompass_Preferred

NBD-556 is a small-molecule CD4 mimetic that inhibits HIV-1 entry by blocking the gp120-CD4 interaction. It is supplied as a solid research compound for use in biochemical and cell-based assays investigating viral entry and fusion.

  • CD4 mimetic that blocks the gp120-CD4 interaction.
  • Inhibits HIV-1 entry and fusion in cell-based assays.
  • High purity (99.6%).
  • Molecular weight 337.84 g·mol-1.
  • CAS number 333353-44-9.
  • Soluble in DMSO (≈33.33 mg/mL); ultrasonic treatment may be required.
  • Supplied as a 10 mg solid; store powder at -20°C for long-term storage.

Catalog No. 50-002-98491 Supplier Medchemexpress LLC Supplier No. HY7664810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NBD-556 is a small-molecule CD4 mimetic that inhibits HIV-1 entry by blocking the gp120-CD4 interaction. It is supplied as a solid research compound for use in biochemical and cell-based assays investigating viral entry and fusion.

  • CD4 mimetic that blocks the gp120-CD4 interaction.
  • Inhibits HIV-1 entry and fusion in cell-based assays.
  • High purity (99.6%).
  • Molecular weight 337.84 g·mol-1.
  • CAS number 333353-44-9.
  • Soluble in DMSO (≈33.33 mg/mL); ultrasonic treatment may be required.
  • Supplied as a 10 mg solid; store powder at -20°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.