Learn More
Medchemexpress LLC Nbd-556 | 333353-44-9 | 99.6% | C17H24ClN3O2 | 1 ML
SDP
Catalog No. 5000434475
Encompass_Preferred
NBD-556, a CD4 mimetic, is a potent HIV-1 entry inhibitor that blocks the gp120-CD4 interaction. It demonstrates potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Potent HIV-1 entry inhibitor
  • Blocks gp120-CD4 interaction
  • Shows potent cell fusion and virus-cell fusion inhibitory activity
  • Inhibits HIV-1 envelope-mediated virus-cell and cell-cell fusion
  • Inhibits infection of MT-2 cells by laboratory-adapted HIV-1 strains
  • Inhibits CD4-dependent virus in a dose-dependent manner
Catalog No. 50-004-34475 Supplier Medchemexpress LLC Supplier No. HY7664810MM/1ML
NBD-556, a CD4 mimetic, is a potent HIV-1 entry inhibitor that blocks the gp120-CD4 interaction. It demonstrates potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Potent HIV-1 entry inhibitor
  • Blocks gp120-CD4 interaction
  • Shows potent cell fusion and virus-cell fusion inhibitory activity
  • Inhibits HIV-1 envelope-mediated virus-cell and cell-cell fusion
  • Inhibits infection of MT-2 cells by laboratory-adapted HIV-1 strains
  • Inhibits CD4-dependent virus in a dose-dependent manner