Learn More
Medchemexpress LLC NBD-556 | 333353-44-9 | 99.6% | C17H24ClN3O2 | 25 MG
SDP
Catalog No. 5000358129
Encompass_Preferred
NBD-556 is a CD4 mimetic and a potent HIV-1 entry inhibitor. It functions by blocking the gp120-CD4 interaction. NBD-556 exhibits potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Inhibits HIV-1 envelope-mediated virus-cell and cell-cell fusion.
  • Inhibits the infection of MT-2 cells by laboratory-adapted HIV-1 strains.
  • Inhibits CD4-dependent virus in a dose-dependent manner.
Catalog No. 50-003-58129 Supplier Medchemexpress LLC Supplier No. 25MGHY76648
NBD-556 is a CD4 mimetic and a potent HIV-1 entry inhibitor. It functions by blocking the gp120-CD4 interaction. NBD-556 exhibits potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Inhibits HIV-1 envelope-mediated virus-cell and cell-cell fusion.
  • Inhibits the infection of MT-2 cells by laboratory-adapted HIV-1 strains.
  • Inhibits CD4-dependent virus in a dose-dependent manner.