Learn More
Selleck Chemical LLC NBD-556-E0943-5MG
SDP
Catalog No. 503136527
Encompass_Preferred
NBD-556 is a novel human immunodeficiency virus type 1 (HIV-1) entry inhibitor that block the gp120-CD4 interaction
Catalog No. 50-313-6527 Supplier Selleck Chemical LLC Supplier No. E09435MG
NBD-556 is a novel human immunodeficiency virus type 1 (HIV-1) entry inhibitor that block the gp120-CD4 interaction