Learn More
Medchemexpress LLC NBD-557 | 333352-59-3 | 99.4% | C17H24BrN3O2 | 1 ML
SDP
Catalog No. 5000434437
Encompass_Preferred
NBD-557 is a potential HIV-1 inhibitor and a small molecule organic compound with drug-like properties. It exhibits potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Targets viral entry by inhibiting the binding of HIV-1 envelope glycoprotein gp120 to the cellular receptor CD4.
  • Active against both X4 and R5 viruses, suggesting its anti-HIV-1 activity is not dependent on coreceptor tropism.
  • Binds to unliganded HIV-1 gp120 but not to the cellular receptor CD4.
  • Active against HIV-1 laboratory-adapted strains, including an AZT-resistant strain, and HIV-1 primary isolates.
Catalog No. 50-004-34437 Supplier Medchemexpress LLC Supplier No. HY7664910MM/1ML
NBD-557 is a potential HIV-1 inhibitor and a small molecule organic compound with drug-like properties. It exhibits potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Targets viral entry by inhibiting the binding of HIV-1 envelope glycoprotein gp120 to the cellular receptor CD4.
  • Active against both X4 and R5 viruses, suggesting its anti-HIV-1 activity is not dependent on coreceptor tropism.
  • Binds to unliganded HIV-1 gp120 but not to the cellular receptor CD4.
  • Active against HIV-1 laboratory-adapted strains, including an AZT-resistant strain, and HIV-1 primary isolates.