Learn More
Medchemexpress LLC NBD-557 | 333352-59-3 | 99.4% | C17H24BrN3O2 | 25 MG
SDP
Catalog No. 5000358238
Encompass_Preferred
NBD-557 is a small molecule organic compound identified as a potential HIV-1 inhibitor. It targets viral entry by inhibiting the binding of HIV-1 envelope glycoprotein gp120 to the CD4 receptor without affecting later stages of the HIV-1 life cycle.
  • Exhibits potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Active against X4 and R5 viruses, and laboratory-adapted HIV-1 strains, including an AZT-resistant strain.
  • Binds to unliganded HIV-1 gp120, but not to the cellular receptor CD4.
Catalog No. 50-003-58238 Supplier Medchemexpress LLC Supplier No. 25MGHY76649
NBD-557 is a small molecule organic compound identified as a potential HIV-1 inhibitor. It targets viral entry by inhibiting the binding of HIV-1 envelope glycoprotein gp120 to the CD4 receptor without affecting later stages of the HIV-1 life cycle.
  • Exhibits potent cell fusion and virus-cell fusion inhibitory activity at low micromolar levels.
  • Active against X4 and R5 viruses, and laboratory-adapted HIV-1 strains, including an AZT-resistant strain.
  • Binds to unliganded HIV-1 gp120, but not to the cellular receptor CD4.