Learn More
Medchemexpress LLC Nbd-amine | 10199-91-4 | 96.7% | 100 MG
SDP
Catalog No. 5000234701
Encompass_Preferred
NBD-amine is a fluorogenic amine that fluoresces weakly in water and strongly in organic solvents, membranes, and other hydrophobic environments. It is used as an environment-sensitive fluorescent probe and labeling reagent in biochemical and biophysical research.
  • Fluorogenic amine that increases fluorescence in nonpolar environments.
  • Useful as a fluorescent probe and labeling reagent for biomolecules.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Available in multiple pack sizes, including 100 mg.
  • Supplied with reported purity suitable for research applications.
Catalog No. 50-002-34701 Supplier Medchemexpress LLC Supplier No. HY119791100MG
NBD-amine is a fluorogenic amine that fluoresces weakly in water and strongly in organic solvents, membranes, and other hydrophobic environments. It is used as an environment-sensitive fluorescent probe and labeling reagent in biochemical and biophysical research.
  • Fluorogenic amine that increases fluorescence in nonpolar environments.
  • Useful as a fluorescent probe and labeling reagent for biomolecules.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Available in multiple pack sizes, including 100 mg.
  • Supplied with reported purity suitable for research applications.