Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NBD-amine | 10199-91-4 | 96.7% | 50 MG
SDP

Catalog No. 5000231868
Encompass_Preferred

NBD-amine is a fluorogenic amine used as a fluorescent probe and reagent. It fluoresces weakly in water and strongly in organic solvents, membranes, and other hydrophobic environments, and is supplied as a powder with demonstrated solubility in DMSO.

  • Fluorogenic amine that increases fluorescence in hydrophobic environments.
  • CAS number 10199-91-4; molecular weight 180.12 g·mol⁻¹; formula C6H4N4O3.
  • Purity 96.7% as supplied.
  • Soluble in DMSO (≈31.25 mg/mL); hygroscopic DMSO affects solubility.
  • Storage: powder -20°C (stable up to 3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple package sizes including 50 MG.

Catalog No. 50-002-31868 Supplier Medchemexpress LLC Supplier No. HY11979150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NBD-amine is a fluorogenic amine used as a fluorescent probe and reagent. It fluoresces weakly in water and strongly in organic solvents, membranes, and other hydrophobic environments, and is supplied as a powder with demonstrated solubility in DMSO.

  • Fluorogenic amine that increases fluorescence in hydrophobic environments.
  • CAS number 10199-91-4; molecular weight 180.12 g·mol⁻¹; formula C6H4N4O3.
  • Purity 96.7% as supplied.
  • Soluble in DMSO (≈31.25 mg/mL); hygroscopic DMSO affects solubility.
  • Storage: powder -20°C (stable up to 3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple package sizes including 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.