Learn More
Apexbio Technology LLC NBD-Cl 10199-89-0 1g
SDP
Catalog No. 5000089842
Encompass_Preferred
NBD-Cl (10199-89-0) is a fluorescent labeling reagent that covalently reacts with nucleophilic groups specifically amines and thiols It is designed to derivatize these functional groups enabling detection through the formation of highly fluorescent conjugates NBD-Cl exerts its biological utility primarily through its electrophilic reaction mechanism with free amino and thiol groups in biomolecules No specific experimental model or quantitative activity metric is provided Based on these chemical properties NBD-Cl holds research potential in the labeling and detection of amino acid residues peptides proteins and thiol-containing enzymes as well as in studies of enzyme activities and biomolecular interactions
Catalog No. 50-000-89842 Supplier Apexbio Technology LLC Supplier No. C65651000
NBD-Cl (10199-89-0) is a fluorescent labeling reagent that covalently reacts with nucleophilic groups specifically amines and thiols It is designed to derivatize these functional groups enabling detection through the formation of highly fluorescent conjugates NBD-Cl exerts its biological utility primarily through its electrophilic reaction mechanism with free amino and thiol groups in biomolecules No specific experimental model or quantitative activity metric is provided Based on these chemical properties NBD-Cl holds research potential in the labeling and detection of amino acid residues peptides proteins and thiol-containing enzymes as well as in studies of enzyme activities and biomolecular interactions