Learn More
Medchemexpress LLC HY-D0042 1g Medchemexpress, NBD-Cl CAS:10199-89-0 Purity:>98%
SDP
Catalog No. 501933139
Encompass_Preferred
Medchemexpress, HY-D0042 1g NBD-Cl CAS:10199-89-0 NBD-Cl is a nonfluorescent reagent which becomes highly fluorescent after reaction with thiol or amino groups. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3139 Supplier Medchemexpress LLC Supplier No. HYD00421G
Medchemexpress, HY-D0042 1g NBD-Cl CAS:10199-89-0 NBD-Cl is a nonfluorescent reagent which becomes highly fluorescent after reaction with thiol or amino groups. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.