Learn More
AdipoGen NBD-Cl
SDP
Supplier:  AdipoGen CDXC0010G050
Encompass_Preferred
Chemical. CAS 10199-89-0. Formula C6H2ClN3O3. MW 199.55. Synthetic. NBD chloride is nonfluorescent until it reacts with primary or secondary amines to produce a fluorescent product. NBD chloride has been extensively used as a derivatizing reagent for chromatography analysis of amino acids and low molecular weight amines.
Catalog No. 50-256-1557