Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NBD-Cl (NBD chloride) | 10199-89-0 | 99.5% | 1 ML
SDP

Catalog No. 5000410035
Encompass_Preferred

NBD-Cl is a nonfluorescent reagent that becomes highly fluorescent after reacting with thiol or amino groups. It forms highly fluorescent derivatives for detecting all protein amino acids and provides a simple and sensitive method for determining N-terminal amino acids. It also allows for the identification of prolyl peptides based on fluorescence characteristics.

  • Nonfluorescent reagent that becomes highly fluorescent after reaction with thiol or amino groups
  • Forms fluorescent derivatives for detection of all protein amino acids
  • Simple and sensitive method for determining N-terminal amino acids
  • Differences in intensity and color of fluorescence can identify prolyl peptides
  • Appearance: Solid, light yellow to yellow
  • Excitation (Ex): 498, Emission (Em): 515
  • Soluble in DMSO (≥ 100 mg/mL)
  • Insoluble in H2O (< 0.1 mg/mL)

Catalog No. 50-004-10035 Supplier Medchemexpress LLC Supplier No. HYD004210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NBD-Cl is a nonfluorescent reagent that becomes highly fluorescent after reacting with thiol or amino groups. It forms highly fluorescent derivatives for detecting all protein amino acids and provides a simple and sensitive method for determining N-terminal amino acids. It also allows for the identification of prolyl peptides based on fluorescence characteristics.

  • Nonfluorescent reagent that becomes highly fluorescent after reaction with thiol or amino groups
  • Forms fluorescent derivatives for detection of all protein amino acids
  • Simple and sensitive method for determining N-terminal amino acids
  • Differences in intensity and color of fluorescence can identify prolyl peptides
  • Appearance: Solid, light yellow to yellow
  • Excitation (Ex): 498, Emission (Em): 515
  • Soluble in DMSO (≥ 100 mg/mL)
  • Insoluble in H2O (< 0.1 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.