Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBD-CO-Hz 95.0+%, TCI America™
SDP

Catalog No. A5573100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5573100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5573100MG Supplier TCI America Supplier No. A5573100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[for HPLC Labeling]

Chemical Identifiers

CAS 221263-97-4
Molecular Formula C9H10N6O4
Molecular Weight (g/mol) 266.217
MDL Number MFCD02093433
InChI Key IDJGYEQXPMQJKF-UHFFFAOYSA-N
Synonym 4-(N-Hydrazinocarbonylmethyl-N-methylamino)-7-nitrobenzofurazan, 4-(N-Hydrazinocarbonylmethyl-N-methylamino)-7-nitro-2,1,3-benzoxadiazole
PubChem CID 3428975
IUPAC Name 2-[methyl-(4-nitro-2,1,3-benzoxadiazol-7-yl)amino]acetohydrazide
SMILES CN(CC(=O)NN)C1=CC=C(C2=NON=C12)[N+](=O)[O-]

Specifications

Quantity 100 mg
Formula Weight 266.22
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material NBD-CO-Hz [=4-(N-Hydrazinocarbonylmethyl-N-methylamino)-7-nitro-2,1,3-benzoxadiazole] [for HP
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.