Learn More
AdipoGen NBD-dodecanoic acid
SDP
Supplier:  AdipoGen CDXN0013M500
Encompass_Preferred
Chemical. CAS 96801-39-7. Formula C18H26N4O5. MW 378.42. Synthetic. Suitable for probing the ligand binding sites of fatty acid and sterol carrier proteins as well as the investigation of lipolysis processes.
Catalog No. 50-256-0439