Learn More
AdipoGen NBD dodecanoic acid N-SE
SDP
Catalog No. 502560393
Encompass_Preferred
Chemical. CAS 689263-76-1. Formula C22H29N5O7. MW 475.5. Synthetic. Suitable for probing the ligand binding sites of fatty acid and sterol carrier proteins as well as the investigation of lipolysis processes.
Catalog No. 50-256-0393 Supplier AdipoGen Supplier No. CDXN0147M025
Chemical. CAS 689263-76-1. Formula C22H29N5O7. MW 475.5. Synthetic. Suitable for probing the ligand binding sites of fatty acid and sterol carrier proteins as well as the investigation of lipolysis processes.