Learn More
Apexbio Technology LLC NBD-F 29270-56-2 100mg
SDP
Catalog No. 5000093982
Encompass_Preferred
NBD-F (29270-56-2) is a fluorescent derivatization reagent utilized for the detection and quantification of amino acids and low molecular weight amines NBD-F exerts its activity by selectively reacting with primary and secondary amines through its benzofurazan structure forming fluorescent conjugates detectable in biological samples Based on these properties NBD-F holds research potential in biochemical analysis metabolic profiling disease pathology investigation and fluorescent labeling in proteomics and molecular biology
Catalog No. 50-000-93982 Supplier Apexbio Technology LLC Supplier No. A4676100
NBD-F (29270-56-2) is a fluorescent derivatization reagent utilized for the detection and quantification of amino acids and low molecular weight amines NBD-F exerts its activity by selectively reacting with primary and secondary amines through its benzofurazan structure forming fluorescent conjugates detectable in biological samples Based on these properties NBD-F holds research potential in biochemical analysis metabolic profiling disease pathology investigation and fluorescent labeling in proteomics and molecular biology