Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NBD-F (4-Fluoro-7-nitrobenzofurazan) | 29270-56-2 | 98.87% | 1 ML
SDP

Catalog No. 5000410495
Encompass_Preferred

NBD-F (4-Fluoro-7-nitrobenzofurazan) is a pro-fluorescent reagent developed for amino acid analysis. It reacts with primary or secondary amines to create a fluorescent product, which is used for the analysis of amino acids and low molecular weight amines. NBD-F is also applied in the analysis of other amino acid derivatives like N-methyl-D-aspartic acid and glutathione. This reagent offers a simple derivatization procedure and its derivatives are highly stable.

  • Pro-fluorescent reagent developed for amino acid analysis.
  • Reacts with primary or secondary amines to produce a fluorescent product.
  • Used for analysis of amino acids and low molecular weight amines, and other amino acid derivatives.
  • Simple derivatization procedure, and derivatives are highly stable.
  • Appearance: solid.
  • Color: light yellow to yellow.
  • Emission (Em): 515.
  • Excitation (Ex): 445.

Catalog No. 50-004-10495 Supplier Medchemexpress LLC Supplier No. HYD078510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NBD-F (4-Fluoro-7-nitrobenzofurazan) is a pro-fluorescent reagent developed for amino acid analysis. It reacts with primary or secondary amines to create a fluorescent product, which is used for the analysis of amino acids and low molecular weight amines. NBD-F is also applied in the analysis of other amino acid derivatives like N-methyl-D-aspartic acid and glutathione. This reagent offers a simple derivatization procedure and its derivatives are highly stable.

  • Pro-fluorescent reagent developed for amino acid analysis.
  • Reacts with primary or secondary amines to produce a fluorescent product.
  • Used for analysis of amino acids and low molecular weight amines, and other amino acid derivatives.
  • Simple derivatization procedure, and derivatives are highly stable.
  • Appearance: solid.
  • Color: light yellow to yellow.
  • Emission (Em): 515.
  • Excitation (Ex): 445.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.