Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NBD-F (4-fluoro-7-nitrobenzofurazan) | 29270-56-2 | MFCD00010196 | 98.0% | 183.10 | C6H2FN3O3 | 10 MG
SDP

Catalog No. 5000297324
Encompass_Preferred

NBD-F (4-fluoro-7-nitrobenzofurazan) is a pro-fluorescent derivatization reagent that reacts with primary and secondary amines to produce stable fluorescent adducts. It is used for amino acid analysis, HPLC fluorescence detection, and other sensitive analytical applications.

  • Pro-fluorescent reagent for derivatizing primary and secondary amines.
  • Excitation 445 nm, emission 515 nm for fluorescence detection.
  • Molecular weight 183.10 g/mol; formula C6H2FN3O3.
  • Typical purity approximately 98.0%.
  • Light yellow solid; store protected from light at 4°C; freeze solutions for long-term storage.

Catalog No. 50-002-97324 Supplier Medchemexpress LLC Supplier No. HYD078510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NBD-F (4-fluoro-7-nitrobenzofurazan) is a pro-fluorescent derivatization reagent that reacts with primary and secondary amines to produce stable fluorescent adducts. It is used for amino acid analysis, HPLC fluorescence detection, and other sensitive analytical applications.

  • Pro-fluorescent reagent for derivatizing primary and secondary amines.
  • Excitation 445 nm, emission 515 nm for fluorescence detection.
  • Molecular weight 183.10 g/mol; formula C6H2FN3O3.
  • Typical purity approximately 98.0%.
  • Light yellow solid; store protected from light at 4°C; freeze solutions for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.