Learn More
Cayman Chemical NBD-Fructose 10mg
SDP
Catalog No. 502892707
Encompass_Preferred
A fluorescent derivative of fructose; has been used to monitor fructose uptake by the GLUT5 transporter in MCF-7, MDA-MB-435, and MDA-MB-231 human breast cancer cells; ex/em = 472/538 nm, respectively
Catalog No. 50-289-2707 Supplier Cayman Chemical Supplier No. 900231410 MG
Add to Cart
Add to Cart
A fluorescent derivative of fructose; has been used to monitor fructose uptake by the GLUT5 transporter in MCF-7, MDA-MB-435, and MDA-MB-231 human breast cancer cells; ex/em = 472/538 nm, respectively