Learn More
Cayman Chemical NBD-Fructose, Purity: ≥95% (sum of isomers), CAS: 940961-04-6, 1 mg
SDP
Catalog No. 502892533
Encompass_Preferred
A fluorescent derivative of fructose; has been used to monitor fructose uptake by the GLUT5 transporter in MCF-7, MDA-MB-435, and MDA-MB-231 human breast cancer cells; ex/em = 472/538 nm, respectively
Catalog No. 50-289-2533 Supplier Cayman Chemical Supplier No. 90023141 MG
A fluorescent derivative of fructose; has been used to monitor fructose uptake by the GLUT5 transporter in MCF-7, MDA-MB-435, and MDA-MB-231 human breast cancer cells; ex/em = 472/538 nm, respectively