Learn More
Cayman Chemical NBD-Fructose 500UG
SDP
Catalog No. 502893657
Encompass_Preferred
A fluorescent derivative of fructose; has been used to monitor fructose uptake by the GLUT5 transporter in MCF-7, MDA-MB-435, and MDA-MB-231 human breast cancer cells; ex/em = 472/538 nm, respectively
Catalog No. 50-289-3657 Supplier Cayman Chemical Supplier No. 9002314500 UG
A fluorescent derivative of fructose; has been used to monitor fructose uptake by the GLUT5 transporter in MCF-7, MDA-MB-435, and MDA-MB-231 human breast cancer cells; ex/em = 472/538 nm, respectively