Learn More
AdipoGen NBD-H
SDP
Supplier:  AdipoGen CDXH0022M100
Encompass_Preferred
Chemical. CAS 90421-78-6. Formula C6H5N5O3. MW 195.14. Synthetic. Fluorogenic reagent for aldehydes and ketones. Used in fluorometric detection of aliphatic aldehydes and for HPLC labeling. lambda ex 445nm, lambda em 522 nm in chloroform after derivatization with formaldehyde. Fluorescent reagent for protein labeling and pre-column derivatization for chromatographic procedures such as HPLC.
Catalog No. 50-256-0800