Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBD-H (=4-Hydrazino-7-nitro-2,1,3-benzoxadiazole Hydrazine) 98.0+%, TCI America™
SDP

Catalog No. A5557100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5557100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5557100MG Supplier TCI America Supplier No. A5557100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 131467-87-3
Molecular Formula C6H9N7O3
Molecular Weight (g/mol) 227.18
MDL Number MFCD00191508
InChI Key BLIOAKMSUDUCKB-UHFFFAOYSA-N
Synonym 4-Hydrazino-7-nitrobenzofurazan Hydrazine, 4-Hydrazino-7-nitro-2,1,3-benzoxadiazole Hydrazine
PubChem CID 16218437
IUPAC Name 4-hydrazinyl-7-nitro-2,1,3-benzoxadiazole; hydrazine
SMILES NN.NNC1=CC=C(C2=NON=C12)[N+]([O-])=O

Specifications

Quantity 100 mg
Formula Weight 227.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material NBD-H (=4-Hydrazino-7-nitro-2,1,3-benzoxadiazole Hydrazine) [for HPLC Labeling]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.