Learn More
Enzo Life Sciences NBD peptide / NF-kappaB blocker (cell permeable) (1Vial)
SDP
Catalog No. 502007848
Encompass_Preferred
For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter. Alternative name: NEMO binding domain peptide. Formula: C121H202N48O32. MW: 2841.2. Purity: ≥95% (HPLC). Formulation: Liquid. In sterile PBS. Long Term Storage: -80°C. Handling: Keep on ice at all times. Protect from moisture. Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
Catalog No. 50-200-7848 Supplier Enzo Life Sciences Supplier No. ALX1630111
For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter. Alternative name: NEMO binding domain peptide. Formula: C121H202N48O32. MW: 2841.2. Purity: ≥95% (HPLC). Formulation: Liquid. In sterile PBS. Long Term Storage: -80°C. Handling: Keep on ice at all times. Protect from moisture. Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.