Learn More
AdipoGen NBD-NMe2
SDP
Catalog No. 502561035
Encompass_Preferred
Chemical. CAS 1455-87-4. Formula C8H8N4O3. MW 208.17. Building block and interemediate for organic synthesis. Fluorescent dye. Derivative of the dipolar fluorescent dye NBD. Spectral data lambdaEx=498, lambdaEm=568, Extinction Coefficient=17,000 M-1 cm-1.
Catalog No. 50-256-1035 Supplier AdipoGen Supplier No. CDXD0834M250
Chemical. CAS 1455-87-4. Formula C8H8N4O3. MW 208.17. Building block and interemediate for organic synthesis. Fluorescent dye. Derivative of the dipolar fluorescent dye NBD. Spectral data lambdaEx=498, lambdaEm=568, Extinction Coefficient=17,000 M-1 cm-1.