Learn More
Medchemexpress LLC NBD-Pen | 1955505-54-0 | 99.9% | 50 MG
SDP
Catalog No. 5000338815
Encompass_Preferred
NBD-Pen is a fluorescent probe designed for the selective and sensitive detection of lipid radicals in biological systems. It functions by turn-on fluorescence to directly detect lipid radicals in living cells.
  • First fluorescence probe for lipid radicals
  • Offers high selectivity and sensitivity (λex: 470 nm, λem: 530 nm)
  • Specifically detects lipid-derived radicals over other reactive species (e.g., H2O2, ClO-, O2-?, and ?OH)
  • Directly detects lipid radicals in living cells through turn-on fluorescence
  • Decreases inflammation, apoptosis, and oxidative stress markers
  • Applicable in various disease models such as hepatic carcinoma
Catalog No. 50-003-38815 Supplier Medchemexpress LLC Supplier No. HY12910950MG
NBD-Pen is a fluorescent probe designed for the selective and sensitive detection of lipid radicals in biological systems. It functions by turn-on fluorescence to directly detect lipid radicals in living cells.
  • First fluorescence probe for lipid radicals
  • Offers high selectivity and sensitivity (λex: 470 nm, λem: 530 nm)
  • Specifically detects lipid-derived radicals over other reactive species (e.g., H2O2, ClO-, O2-?, and ?OH)
  • Directly detects lipid radicals in living cells through turn-on fluorescence
  • Decreases inflammation, apoptosis, and oxidative stress markers
  • Applicable in various disease models such as hepatic carcinoma