Learn More
Medchemexpress LLC NBD-Pen | 1955505-54-0 | 99.94% | 10 MG
SDP
Catalog No. 5000338756
Encompass_Preferred
NBD-Pen is the first fluorescence probe for lipid radicals, offering high selectivity and sensitivity (λex: 470 nm, λem: 530 nm). It specifically detects lipid-derived radicals over other reactive species found in biological systems, including H2O2, ClO-, O2•-, and •OH. NBD-Pen directly detects lipid radicals in living cells through turn-on fluorescence. It has been observed to decrease inflammation, apoptosis, and oxidative stress markers, and can be utilized in various disease models such as hepatic carcinoma.
  • First fluorescence probe for lipid radicals.
  • High selectivity and sensitivity (λex: 470 nm, λem: 530 nm).
  • Specifically detects lipid-derived radicals over other reactive species (H2O2, ClO-, O2•-, and •OH).
  • Directly detects lipid radicals in living cells by turn-on fluorescence.
  • Decreases inflammation, apoptosis, and oxidative stress markers.
  • Applicable for studying various disease models, including hepatic carcinoma.
Catalog No. 50-003-38756 Supplier Medchemexpress LLC Supplier No. HY12910910MG
NBD-Pen is the first fluorescence probe for lipid radicals, offering high selectivity and sensitivity (λex: 470 nm, λem: 530 nm). It specifically detects lipid-derived radicals over other reactive species found in biological systems, including H2O2, ClO-, O2•-, and •OH. NBD-Pen directly detects lipid radicals in living cells through turn-on fluorescence. It has been observed to decrease inflammation, apoptosis, and oxidative stress markers, and can be utilized in various disease models such as hepatic carcinoma.
  • First fluorescence probe for lipid radicals.
  • High selectivity and sensitivity (λex: 470 nm, λem: 530 nm).
  • Specifically detects lipid-derived radicals over other reactive species (H2O2, ClO-, O2•-, and •OH).
  • Directly detects lipid radicals in living cells by turn-on fluorescence.
  • Decreases inflammation, apoptosis, and oxidative stress markers.
  • Applicable for studying various disease models, including hepatic carcinoma.