Learn More
AdipoGen NBD-PZ
SDP
Supplier:  AdipoGen CDXN0014G001
Encompass_Preferred
Chemical. CAS 139332-66-4. Formula C10H11N5O3. MW 249.23. Synthetic. Fluorescent reagent for the determination of mono- and diisocyantes in air samples, and determination of lactate, hydroxybutyrate and other intermediates in body fluids. The piperazino moiety reacts readily with isocyantes to form the corresponding urea-derivatives. The derivatives can be detected by LC with UV or fluorescent-detection. Used for visualizing lysosomes in living cells also.
Catalog No. 50-256-0438