Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBD-PZ (=4-Nitro-7-piperazino-2,1,3-benzoxadiazole) 98.0+%, TCI America™
SDP

Catalog No. A5554100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5554100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5554100MG Supplier TCI America Supplier No. A5554100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 139332-66-4
Molecular Formula C10H11N5O3
Molecular Weight (g/mol) 249.23
MDL Number MFCD00191507
InChI Key MVLWYDGJBGPXOL-UHFFFAOYSA-N
Synonym 4-Nitro-7-piperazinobenzofurazan, 4-Nitro-7-piperazino-2,1,3-benzoxadiazole
PubChem CID 10586684
IUPAC Name 4-nitro-7-(piperazin-1-yl)-2,1,3-benzoxadiazole
SMILES [O-][N+](=O)C1=CC=C(N2CCNCC2)C2=NON=C12

Specifications

Melting Point 210°C
Quantity 100 mg
Formula Weight 249.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material NBD-PZ (=4-Nitro-7-piperazino-2,1,3-benzoxadiazole) [for HPLC Labeling]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.