Learn More
AdipoGen NBD-X
SDP
Catalog No. 502560445
Encompass_Preferred
Chemical. CAS 88235-25-0. Formula C12H14N4O5. MW 294.26. Synthetic. Fluorescent probe used for investigation of binding sites of fatty acid and sterol carrier proteins, also used for cell membrane staining.
Catalog No. 50-256-0445 Supplier AdipoGen Supplier No. CDXN0005G001
Chemical. CAS 88235-25-0. Formula C12H14N4O5. MW 294.26. Synthetic. Fluorescent probe used for investigation of binding sites of fatty acid and sterol carrier proteins, also used for cell membrane staining.