Learn More
Medchemexpress LLC NBD-X acid | 88235-25-0 | 99.18% | 50 MG
SDP
Catalog No. 5000410009
Encompass_Preferred
NBD-X acid is a fluorescent probe containing a nitrobenzoxadiazole group. It can be used to study the structure and function of biomolecules.
  • Fluorescent probe containing a nitrobenzoxadiazole group.
  • Useful for studying the structure and function of biomolecules.
Catalog No. 50-004-10009 Supplier Medchemexpress LLC Supplier No. HYD013350MG
NBD-X acid is a fluorescent probe containing a nitrobenzoxadiazole group. It can be used to study the structure and function of biomolecules.
  • Fluorescent probe containing a nitrobenzoxadiazole group.
  • Useful for studying the structure and function of biomolecules.