Learn More
AdipoGen NBD-X SE
SDP
Catalog No. 502560432
Encompass_Preferred
Chemical. CAS 145195-58-0. Formula C16H17N5O7. MW 391.33. Synthetic. NBD-X, SE is an excellent amino-reactive building block that can be used to prepare peptide conjugates and other bioconjugates/biopolymers. It has much higher conjugation efficiency than NBD-Cl and NBD-F. This amine-reactive NBD derivative, containing a convenient succinimidyl ester, can be used to develop environmentally sensitive bioconjugates. NBD is a functional analog of the dinitrophenyl hapten, and its fluorescence is quenched upon binding to anti-dinitrophenyl anti-DNP antibodies. The X-spacer separates the fluorescent dye from the protein allowing greater sensitivity and reduced perturbation effects on protein labeling. Spectral properties lamdaEx=460 lambdaEm=540.
Catalog No. 50-256-0432 Supplier AdipoGen Supplier No. CDXN0045M025
Chemical. CAS 145195-58-0. Formula C16H17N5O7. MW 391.33. Synthetic. NBD-X, SE is an excellent amino-reactive building block that can be used to prepare peptide conjugates and other bioconjugates/biopolymers. It has much higher conjugation efficiency than NBD-Cl and NBD-F. This amine-reactive NBD derivative, containing a convenient succinimidyl ester, can be used to develop environmentally sensitive bioconjugates. NBD is a functional analog of the dinitrophenyl hapten, and its fluorescence is quenched upon binding to anti-dinitrophenyl anti-DNP antibodies. The X-spacer separates the fluorescent dye from the protein allowing greater sensitivity and reduced perturbation effects on protein labeling. Spectral properties lamdaEx=460 lambdaEm=540.