Learn More
Abcam NBD- x , SE, 25MG
Catalog No. 50996111
Encompass_Preferred
MW 391.34 Da, Purity >90%. Lipophilic green fluorescent probe for labeling antibodies and proteins. X-spacer provides separation between dye and protein for increased sensitivity and reduced impact on protein function. Functional analog of the dinitrophenol (DNP) hapten. Fluorescence is quenched when bound by anti-DNP antibodies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-111 Supplier Abcam Supplier No. AB27505525MG
MW 391.34 Da, Purity >90%. Lipophilic green fluorescent probe for labeling antibodies and proteins. X-spacer provides separation between dye and protein for increased sensitivity and reduced impact on protein function. Functional analog of the dinitrophenol (DNP) hapten. Fluorescence is quenched when bound by anti-DNP antibodies.
The product is subject to the following: Abcam Restricted Use Statement