Learn More
Bon Opus Biosciences Recombinant Human Neurobeachin/NBEA - Source: E. coli
SDP
Catalog No. NC2733670
Encompass_Preferred
Recombinant Human Neurobeachin/NBEA is produced by E.coli expression system and the target gene encoding T1589-L1703 is expressed. Accession#Q8NFP9. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733670 Supplier Bon Opus Biosciences Supplier No. BP232750UG
Recombinant Human Neurobeachin/NBEA is produced by E.coli expression system and the target gene encoding T1589-L1703 is expressed. Accession#Q8NFP9. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4