Learn More
Aviva Systems Biology Corp NBEAL1 Antibody - N-terminal region (ARP41207_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41207_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NBEAL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NBEAL1. Alternate names: A530083I02Rik, ALS2CR17, ALS2CR16ApplicationWB. Species reactivity: Dog, Guinea Pig, Human
Catalog No. 50-014-57760