Learn More
ABclonal Technology NBEAL2 Rabbit pAb
SDP
Catalog No. 5000105909
Encompass_Preferred
The protein encoded by this gene contains a beige and Chediak-Higashi (BEACH) domain and multiple WD40 domains, and may play a role in megakaryocyte alpha-granule biogenesis. Mutations in this gene are a cause of gray platelet syndrome.
Catalog No. 50-001-05909 Supplier ABclonal Technology Supplier No. A157971000UL
The protein encoded by this gene contains a beige and Chediak-Higashi (BEACH) domain and multiple WD40 domains, and may play a role in megakaryocyte alpha-granule biogenesis. Mutations in this gene are a cause of gray platelet syndrome.