Learn More
Cayman Chemical N-Benzylacetamidine (hydrobromide), Purity: ≥98%, CAS: 186545-76-6, 50 mg
SDP
Catalog No. 502955103
Encompass_Preferred
A potent inhibitor of iNOS (IC50 = 0.20 μM), with over 1,000-fold selectivity compared to eNOS (IC50 = 350 μM)
Catalog No. 50-295-5103 Supplier Cayman Chemical Supplier No. 1357050 MG
A potent inhibitor of iNOS (IC50 = 0.20 μM), with over 1,000-fold selectivity compared to eNOS (IC50 = 350 μM)