Learn More
Cayman Chemical NBenzylACTAmIdInhydrobro 100mg
SDP
Catalog No. 502955372
Encompass_Preferred
A potent inhibitor of iNOS (IC50 = 0.20 μM), with over 1,000-fold selectivity compared to eNOS (IC50 = 350 μM)
Catalog No. 50-295-5372 Supplier Cayman Chemical Supplier No. 13570100 MG
A potent inhibitor of iNOS (IC50 = 0.20 μM), with over 1,000-fold selectivity compared to eNOS (IC50 = 350 μM)