Learn More
Medchemexpress LLC NBI-31772 | 374620-70-9 | 98.88% | 341.27 | 10 MG
SDP
Catalog No. 5000344000
Encompass_Preferred
NBI-31772 is a non-selective IGFBP inhibitor with a Ki of 47 nM. It demonstrates neuroprotective effects, reduces infarct volume during cerebral ischemia, and can restore or enhance proteoglycan synthesis in osteoarticular chondrocytes. It is suitable for various research applications.
  • Inhibits IGFBP with a Ki of 47 nM
  • Demonstrates neuroprotective effects
  • Reduces infarct volume during cerebral ischemia
  • Restores or enhances proteoglycan synthesis in osteoarticular chondrocytes
  • Increases the effect of IGF3 on zebrafish oocyte maturation
  • For research use only
Catalog No. 50-003-44000 Supplier Medchemexpress LLC Supplier No. HY110135100MG
NBI-31772 is a non-selective IGFBP inhibitor with a Ki of 47 nM. It demonstrates neuroprotective effects, reduces infarct volume during cerebral ischemia, and can restore or enhance proteoglycan synthesis in osteoarticular chondrocytes. It is suitable for various research applications.
  • Inhibits IGFBP with a Ki of 47 nM
  • Demonstrates neuroprotective effects
  • Reduces infarct volume during cerebral ischemia
  • Restores or enhances proteoglycan synthesis in osteoarticular chondrocytes
  • Increases the effect of IGF3 on zebrafish oocyte maturation
  • For research use only