Learn More
Medchemexpress LLC NBI-31772 hydrate | 00-00-0 | MFCD20274527 | 99.4% | 372.82 g/mol | C17H11NO7 · 7/4 H2O | 5 MG
SDP
Catalog No. 5000263705
Encompass_Preferred
NBI-31772 hydrate is a research small molecule that inhibits interactions between insulin-like growth factor (IGF) and IGF-binding proteins (IGFBPs). It mobilizes bioactive IGF-I from IGF-IGFBP complexes and has been used in preclinical studies demonstrating neuroprotective, anxiolytic, and antidepressant-like effects.
  • Potent inhibitor of IGF-IGFBP interaction.
  • Releases bioactive IGF-I from IGF-IGFBP complexes.
  • Used in neuroprotection and behavioral pharmacology studies.
  • High purity (≈99.4%).
  • Hydrate form with molecular weight 372.82 g/mol.
  • Supplied as a 5 mg research quantity.
Catalog No. 50-002-63705 Supplier Medchemexpress LLC Supplier No. HY110135A5MG
NBI-31772 hydrate is a research small molecule that inhibits interactions between insulin-like growth factor (IGF) and IGF-binding proteins (IGFBPs). It mobilizes bioactive IGF-I from IGF-IGFBP complexes and has been used in preclinical studies demonstrating neuroprotective, anxiolytic, and antidepressant-like effects.
  • Potent inhibitor of IGF-IGFBP interaction.
  • Releases bioactive IGF-I from IGF-IGFBP complexes.
  • Used in neuroprotection and behavioral pharmacology studies.
  • High purity (≈99.4%).
  • Hydrate form with molecular weight 372.82 g/mol.
  • Supplied as a 5 mg research quantity.