Learn More
Cayman Chemical NBI 35965mesylate 10mg
SDP
Catalog No. 502967689
Encompass_Preferred
A selective CRF1 antagonist (Ki = 4 nM); inhibits cAMP accumulation and ACTH production in vitro (pIC50s = 7.1 and 6.9, respectively) and reduces CRF- and stress-induced ACTH production in vivo
Catalog No. 50-296-7689 Supplier Cayman Chemical Supplier No. 1852410 MG
A selective CRF1 antagonist (Ki = 4 nM); inhibits cAMP accumulation and ACTH production in vitro (pIC50s = 7.1 and 6.9, respectively) and reduces CRF- and stress-induced ACTH production in vivo