Learn More
Medchemexpress LLC HY-15320 5mg Medchemexpress, NBI-74330 CAS:855527-92-3 Purity:>98%
SDP
Catalog No. 502081705
Encompass_Preferred
Medchemexpress, HY-15320 5mg NBI-74330 CAS:855527-92-3 NBI-74330 is a potent antagonist for CXCR3, and exhibits potent inhibition of (125I)CXCL10 and (125I)CXCL11 specific binding with Ki of 1.5 and 3.2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1705 Supplier Medchemexpress LLC Supplier No. HY153205MG
Medchemexpress, HY-15320 5mg NBI-74330 CAS:855527-92-3 NBI-74330 is a potent antagonist for CXCR3, and exhibits potent inhibition of (125I)CXCL10 and (125I)CXCL11 specific binding with Ki of 1.5 and 3.2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.