Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NBI-961 (formerly CMP3a) | 2225902-98-5 | 99.7% | 568.61 g/mol | C28H27F3N6O2S | 10 MG
SDP

Catalog No. 5000288090
Encompass_Preferred

NBI-961 is a potent NEK2 inhibitor that inhibits proteasomal degradation and induces G2/mitosis arrest and apoptosis in diffuse large B cell lymphoma (DLBCL) cells. It is supplied as a solid, light yellow to yellow research-use-only compound and is available in small amounts for laboratory studies.

  • High purity (99.7%).
  • Molecular weight 568.61 g/mol.
  • Chemical formula C28H27F3N6O2S.
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic).
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • Intended for laboratory research use only; not for human or animal use.

Catalog No. 50-002-88090 Supplier Medchemexpress LLC Supplier No. HY15643710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NBI-961 is a potent NEK2 inhibitor that inhibits proteasomal degradation and induces G2/mitosis arrest and apoptosis in diffuse large B cell lymphoma (DLBCL) cells. It is supplied as a solid, light yellow to yellow research-use-only compound and is available in small amounts for laboratory studies.

  • High purity (99.7%).
  • Molecular weight 568.61 g/mol.
  • Chemical formula C28H27F3N6O2S.
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic).
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • Intended for laboratory research use only; not for human or animal use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.